Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004044.6 | NP_004035.2 | 16 | 1942 | ||
GenBank mRNA | AB062403.1 | - | 16 | 1790 | ||
AK290067.1 | - | 15 | 1924 | |||
AK298142.1 | - | 16 | 2055 | |||
AK313066.1 | - | 16 | 1836 | |||
BC008879.2 | - | 16 | 1785 | 4300570 | ||
D82348.1 | - | 16 | 1830 | |||
D89976.1 | - | 16 | 1812 | |||
EU794654.1 | - | 16 | 1929 | |||
U37436.1 | - | 15 | 1752 |