Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATOH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005172.1 | NP_005163.1 | 1 | 876 | ||
GenBank mRNA | BC069145.1 | - | 1 | 877 | 7217005 | |
BC069578.1 | - | 1 | 871 | |||
BC069594.1 | - | 1 | 876 | 7262394 | ||
BC069604.1 | - | 1 | 876 | 7262406 | ||
BC113623.1 | - | 1 | 912 | 8322675 | ||
BC113625.1 | - | 1 | 912 | 8322677 |