Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000702.3 | NP_000693.1 | 11 | 1559 | ||
GenBank mRNA | AB018321.2 | - | 11 | 1516 | ||
AK091617.1 | - | 11 | 1530 | |||
AK124581.1 | - | 9 | 1231 | |||
AK126573.1 | - | 4 | 559 | |||
AK295729.1 | - | 4 | 536 | |||
AK308773.1 | - | 3 | 666 | |||
AK314296.1 | - | 11 | 1530 | |||
AL831991.1 | - | 10 | 1348 | |||
AL831997.1 | - | 10 | 1374 | |||
BC052271.1 | - | 11 | 1510 | 6672133 | ||
KU177927.1 | - | 10 | 1427 |