Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000702.3 | NP_000693.1 | 19 | 2802 | ||
GenBank mRNA | AB018321.2 | - | 19 | 2759 | ||
AK091617.1 | - | 19 | 2773 | |||
AK124581.1 | - | 17 | 2474 | |||
AK126573.1 | - | 12 | 1751 | |||
AK295729.1 | - | 12 | 1779 | |||
AK299460.1 | - | 4 | 553 | |||
AK314296.1 | - | 19 | 2773 | |||
AL831991.1 | - | 18 | 2591 | |||
AL831997.1 | - | 18 | 2617 | |||
BC052271.1 | - | 19 | 2753 | 6672133 | ||
KU177927.1 | - | 18 | 2670 |