Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001396.2 | NP_001001396.1 | 20, 21 | 4194 | ||
NM_001684.4 | NP_001675.3 | 20, 21 | 4194 | |||
GenBank mRNA | AK026443.1 | - | 1220 | |||
AK292458.1 | - | 19, 20 | 3533 | |||
AK307104.1 | - | 3, 4 | 339 | |||
BC140774.1 | - | 20, 21 | 3866 | 9021642 | ||
BC150293.1 | - | 19, 20 | 3551 | 8860318 | ||
BX537444.1 | - | 20, 21 | 4421 | |||
BX647701.1 | - | 1, 2 | 92 | |||
CR749393.1 | - | 20, 21 | 4124 | |||
CR936708.1 | - | 20, 21 | 4149 | |||
DQ201780.1 | - | 3, 4 | 372 | |||
M25874.1 | - | 19, 20 | 3694 | |||
M83363.1 | - | 19, 20 | 3694 | |||
U42026.1 | - | 3, 4 | 321 | |||
U42061.1 | - | 3, 4 | 321 | |||
U42062.1 | - | 2, 3 | 213 | |||
U42378.1 | - | 3, 4 | 321 |