Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001935.2 | NP_001001935.1 | 3, 4 | 506 | ||
NM_001001937.1 | NP_001001937.1 | 4, 5 | 456 | |||
NM_001257334.1 | NP_001244263.1 | 3, 4 | 433 | |||
NM_001257335.1 | NP_001244264.1 | 3, 4 | 769 | |||
NM_004046.5 | NP_004037.1 | 3, 4 | 433 | |||
XM_017025789.1 | XP_016881278.1 | 3, 4 | 375 | |||
GenBank mRNA | AK092735.1 | - | 3, 4 | 448 | ||
AK098765.1 | - | 3, 4 | 365 | |||
AK129739.1 | - | 3, 4 | 376 | |||
AK129900.1 | - | 3, 4 | 376 | |||
AK289457.1 | - | 3, 4 | 702 | |||
AK294804.1 | - | 1, 2 | 519 | |||
AK302272.1 | - | 3, 4 | 376 | |||
AK310051.1 | - | 3, 4 | 731 | |||
AU120644.1 | - | 3, 4 | 401 | |||
BC011384.1 | - | 3, 4 | 364 | 3867375 | ||
BC016046.1 | - | 3, 4 | 351 | 4646761 | ||
BC019310.1 | - | 3, 4 | 364 | 2823663 | ||
BC028335.1 | - | 3, 4 | 353 | |||
BC039135.1 | - | 3, 4 | 346 | 3873390 | ||
BC042694.1 | - | 3, 4 | 346 | |||
BC064562.1 | - | 3, 4 | 364 | 5582043 | ||
BC067385.1 | - | 3, 4 | 376 | 5491013 | ||
BT007209.1 | - | 3, 4 | 311 | |||
CD244623.1 | - | 3, 4 | 392 | |||
CK002707.1 | - | 3, 4 | 434 | |||
D14710.1 | - | 3, 4 | 374 | |||
FJ224309.1 | - | 3, 4 | 369 | |||
X59066.1 | - | 3, 4 | 369 | |||
X65460.1 | - | 3, 4 | 325 |