Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001697.2 | NP_001688.1 | 4 | 311 | ||
GenBank mRNA | AF088071.1 | - | 2 | 106 | ||
AK124187.1 | - | 3 | 1452 | |||
AK222608.1 | - | 4 | 243 | |||
AK222962.1 | - | 4 | 238 | |||
AK311796.1 | - | 4 | 238 | |||
AV734214.1 | - | 4 | 311 | |||
BC021233.2 | - | 4 | 224 | 4126036 | ||
BC022865.1 | - | 4 | 242 | 4103001 | ||
BT019836.1 | - | 4 | 220 | |||
CD174837.1 | - | 4 | 281 | |||
CR456822.1 | - | 4 | 220 | |||
X83218.1 | - | 4 | 256 |