Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294335.1 | NP_001281264.1 | 33 | 4997 | ||
NM_001294336.1 | NP_001281265.1 | 32 | 4898 | |||
XM_011541848.2 | XP_011540150.1 | 32 | 4954 | |||
XM_011541849.2 | XP_011540151.1 | 31 | 4789 | |||
XM_011541858.2 | XP_011540160.1 | 30 | 3953 | |||
XM_017001899.1 | XP_016857388.1 | 32 | 5084 | |||
XM_017001900.1 | XP_016857389.1 | 31 | 5053 | |||
XM_017001901.1 | XP_016857390.1 | 30 | 4999 | |||
XM_017001902.1 | XP_016857391.1 | 30 | 4997 | |||
XM_017001903.1 | XP_016857392.1 | 30 | 4988 | |||
XM_017001904.1 | XP_016857393.1 | 30 | 4954 | |||
XM_017001905.1 | XP_016857394.1 | 30 | 4953 | |||
XM_017001906.1 | XP_016857395.1 | 29 | 4898 | |||
XM_017001907.1 | XP_016857396.1 | 30 | 4888 | |||
XM_017001908.1 | XP_016857397.1 | 30 | 4889 | |||
XM_017001909.1 | XP_016857398.1 | 29 | 4833 | |||
XM_017001910.1 | XP_016857399.1 | 29 | 4789 | |||
XM_017001911.1 | XP_016857400.1 | 28 | 4733 | |||
XM_017001912.1 | XP_016857401.1 | 29 | 4722 | |||
GenBank mRNA | AB005298.1 | - | 33 | 4969 | ||
AK123392.1 | - | 10 | 2506 | |||
AK296502.1 | - | 26 | 3476 | |||
BC009035.1 | - | 5 | 756 | |||
BC136533.1 | - | 30 | 4566 | 9020523 |
Set Membership: |
G-protein coupled receptor |