Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000465.3 | NP_000456.2 | 8 | 1906 | ||
NM_001282543.1 | NP_001269472.1 | 7 | 1849 | |||
NM_001282545.1 | NP_001269474.1 | 4 | 553 | |||
NM_001282548.1 | NP_001269477.1 | 3 | 496 | |||
NR_104212.1 | 7 | 1757 | ||||
NR_104215.1 | 6 | 1700 | ||||
NR_104216.1 | 7 | 956 | ||||
XM_017004613.1 | XP_016860102.1 | 9 | 1954 | |||
XM_017004614.1 | XP_016860103.1 | 9 | 1954 | |||
GenBank mRNA | AK301843.1 | - | 7 | 1719 | ||
AK314260.1 | - | 8 | 1866 | |||
AX794378.1 | - | 8 | 1837 | |||
BC126426.1 | - | 8 | 1834 | 8992142 | ||
BC126428.1 | - | 8 | 1834 | 8992144 | ||
BX098964.1 | - | 3 | 288 | |||
JF790280.1 | - | 7 | 1780 | |||
JF790281.1 | - | 6 | 1631 | |||
JF790282.1 | - | 7 | 887 | |||
JF790283.1 | - | 4 | 439 | |||
JF790284.1 | - | 3 | 427 | |||
U76638.1 | - | 8 | 1837 |