Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000465.3 | NP_000456.2 | 1, 2 | 296 | ||
NM_001282545.1 | NP_001269474.1 | 1, 2 | 296 | |||
NM_001282549.1 | NP_001269478.1 | 1, 2 | 296 | |||
NR_104212.1 | 1, 2 | 296 | ||||
NR_104216.1 | 1, 2 | 296 | ||||
XM_017004613.1 | XP_016860102.1 | 1, 2 | 245 | |||
XM_017004614.1 | XP_016860103.1 | 1, 2 | 245 | |||
GenBank mRNA | AK223409.1 | - | 1, 2 | 290 | ||
AK301843.1 | - | 1, 2 | 258 | |||
AK314260.1 | - | 1, 2 | 256 | |||
AX794378.1 | - | 1, 2 | 227 | |||
BC126426.1 | - | 1, 2 | 224 | 8992142 | ||
BC126428.1 | - | 1, 2 | 224 | 8992144 | ||
DB088853.1 | - | 1, 2 | 297 | |||
JF790282.1 | - | 1, 2 | 227 | |||
JF790283.1 | - | 1, 2 | 182 | |||
JF790285.1 | - | 1, 2 | 182 | |||
JF790287.1 | - | 1, 2 | 182 | |||
U76638.1 | - | 1, 2 | 227 |