Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199839.1 | NP_001186768.1 | 3 | 230 | ||
NM_004050.4 | NP_004041.1 | 3 | 275 | |||
GenBank mRNA | AK289519.1 | - | 3 | 230 | ||
BC021198.2 | - | 2 | 212 | 3944307 | ||
BC104789.1 | - | 3 | 211 | 8143792 | ||
BC113522.1 | - | 3 | 211 | 8322574 | ||
BP216712.1 | - | 3 | 275 | |||
BT019549.1 | - | 1 | 46 | |||
D87461.1 | - | 3 | 222 | |||
DA196433.1 | - | 3 | 234 | |||
DA288957.1 | - | 3 | 230 | |||
DA451936.1 | - | 3 | 187 | |||
U59747.1 | - | 1 | 46 |
Gene Symbol: BCL2L2-PABPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199864.1 | NP_001186793.1 | 3 | 275 | ||
GenBank mRNA | BC011637.1 | - | 3 | 211 |