Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BFSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161705.1 | NP_001155177.1 | 8 | 1640 | ||
NM_001195.4 | NP_001186.1 | 8 | 1784 | |||
NM_001278606.1 | NP_001265535.1 | 9 | 1775 | |||
NM_001278607.1 | NP_001265536.1 | 8 | 1636 | |||
NM_001278608.1 | NP_001265537.1 | 10 | 1761 | |||
XM_017028005.1 | XP_016883494.1 | 7 | 1699 | |||
GenBank mRNA | AK304673.1 | - | 10 | 1761 | ||
AK316247.1 | - | 9 | 1653 | |||
Y16717.2 | - | 8 | 1744 |