Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001713.2 | NP_001704.2 | 1, 2 | 141 | ||
GenBank mRNA | AK298308.1 | - | 1, 2 | 113 | ||
AK303780.1 | - | 1, 2 | 113 | |||
AK315224.1 | - | 1, 2 | 113 | |||
BC012616.1 | - | 1, 2 | 117 | 4045543 | ||
DA370221.1 | - | 1, 2 | 142 | |||
EU794592.1 | - | 1, 2 | 141 | |||
U50929.1 | - | 1, 2 | 62 |