Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001719.2 | NP_001710.1 | 6 | 1629 | ||
GenBank mRNA | AK094784.1 | - | 2 | 369 | ||
AK291186.1 | - | 6 | 1205 | |||
AK312419.1 | - | 6 | 1202 | |||
BC008584.1 | - | 6 | 1141 | 4183402 | ||
EU679366.1 | - | 6 | 1013 | |||
KJ875974.1 | - | 3 | 224 | |||
M60316.1 | - | 6 | 1196 | |||
X51801.1 | - | 6 | 1222 |