Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001719.2 | NP_001710.1 | 2, 3 | 1141 | ||
GenBank mRNA | AK291186.1 | - | 2, 3 | 717 | ||
AK312419.1 | - | 2, 3 | 714 | |||
BC008584.1 | - | 2, 3 | 653 | 4183402 | ||
EU679366.1 | - | 2, 3 | 525 | |||
M60316.1 | - | 2, 3 | 708 | |||
X51801.1 | - | 2, 3 | 734 |