Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204.6 | NP_001195.2 | 8 | 2216 | ||
XM_011511687.1 | XP_011509989.1 | 8 | 2233 | |||
GenBank mRNA | BC052985.2 | - | 8 | 1642 | 6149698 | |
D50516.1 | - | 8 | 1476 | |||
U20165.1 | - | 8 | 1104 | |||
Z48923.1 | - | 8 | 1449 |