Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302777.1 | NP_001289706.1 | 7 | 1349 | ||
NM_004332.3 | NP_004323.2 | 6 | 1121 | |||
NR_026648.2 | 8 | 1417 | ||||
NR_026649.2 | 7 | 1189 | ||||
GenBank mRNA | AJ617684.1 | - | 6 | 769 | ||
AK297518.1 | - | 7 | 873 | |||
AK314909.1 | - | 7 | 1003 | |||
AL832502.1 | - | 6 | 3116 | |||
BC037778.1 | - | 8 | 1422 | 4799692 | ||
BC106901.1 | - | 6 | 836 | 40031474 | ||
X57653.1 | - | 2 | 213 | |||
X81372.1 | - | 7 | 917 |