Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286818.1 | NP_001273747.1 | 4 | 1278 | ||
NM_001730.4 | NP_001721.2 | 4 | 1892 | |||
GenBank mRNA | AB030824.1 | - | 4 | 1558 | ||
AF132818.1 | - | 4 | 1892 | |||
AF287272.1 | - | 4 | 1667 | |||
AK131397.1 | - | 1 | 1283 | |||
AK296900.1 | - | 5 | 1622 | |||
AK302280.1 | - | 4 | 1278 | |||
AK304284.1 | - | 5 | 1586 | |||
BC007695.1 | - | 4 | 1670 | 3956532 | ||
BC042131.1 | - | 4 | 1529 | 5454169 | ||
BT006641.1 | - | 4 | 1356 | |||
D14520.1 | - | 4 | 1200 | |||
EF208215.1 | - | 4 | 1278 | |||
HF546203.1 | - | 4 | 1157 | |||
HF546204.1 | - | 5 | 943 | |||
HQ628639.1 | - | 5 | 1165 | |||
HQ628640.1 | - | 5 | 1309 | |||
HQ628641.1 | - | 5 | 1335 |