Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000063.5 | NP_000054.2 | 16 | 2255 | ||
NM_001145903.2 | NP_001139375.1 | 14 | 1859 | |||
NM_001178063.2 | NP_001171534.1 | 12 | 1405 | |||
NM_001282457.1 | NP_001269386.1 | 12 | 1462 | |||
NM_001282458.1 | NP_001269387.1 | 16 | 2283 | |||
GenBank mRNA | AK096258.1 | - | 13 | 1560 | ||
AK222537.1 | - | 16 | 2007 | |||
AK222783.1 | - | 16 | 1979 | |||
AK298311.1 | - | 12 | 1405 | |||
AK298808.1 | - | 12 | 1462 | |||
AK300892.1 | - | 14 | 1619 | |||
AK300930.1 | - | 16 | 2021 | |||
AK312581.1 | - | 16 | 2089 | |||
AW515583.1 | - | 3 | 320 | |||
BC043484.1 | - | 16 | 1990 | 5763774 | ||
BX537504.1 | - | 10 | 1992 | |||
X04481.1 | - | 16 | 2015 |