Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000065.3 | NP_000056.2 | 12 | 1969 | ||
NM_001115131.2 | NP_001108603.2 | 12 | 1996 | |||
XM_005248357.2 | XP_005248414.1 | 12 | 2044 | |||
XM_006714496.3 | XP_006714559.1 | 13 | 2101 | |||
XM_011514114.2 | XP_011512416.1 | 13 | 2101 | |||
XM_011514115.2 | XP_011512417.1 | 13 | 2022 | |||
XM_011514116.2 | XP_011512418.1 | 14 | 2138 | |||
XM_011514117.2 | XP_011512419.1 | 14 | 2180 | |||
XM_011514118.2 | XP_011512420.1 | 13 | 1918 | |||
XM_011514119.2 | XP_011512421.1 | 12 | 2044 | |||
XM_011514121.2 | XP_011512423.1 | 8 | 1187 | |||
XM_017009818.1 | XP_016865307.1 | 14 | 2161 | |||
XM_017009819.1 | XP_016865308.1 | 9 | 1364 | |||
GenBank mRNA | AB126592.1 | - | 11 | 1731 | ||
AK292509.1 | - | 12 | 1867 | |||
AK313673.1 | - | 12 | 1888 | |||
BC035723.1 | - | 12 | 1996 | 5591446 | ||
J05024.1 | - | 12 | 1788 | |||
J05064.1 | - | 12 | 1886 |