Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000066.3 | NP_000057.2 | 12 | 1829 | ||
NM_001278543.1 | NP_001265472.1 | 13 | 2051 | |||
NM_001278544.1 | NP_001265473.1 | 13 | 1958 | |||
XM_017002235.1 | XP_016857724.1 | 13 | 1897 | |||
GenBank mRNA | AK292740.1 | - | 12 | 1684 | ||
AK298425.1 | - | 13 | 1875 | |||
AK298444.1 | - | 13 | 2051 | |||
AK313382.1 | - | 12 | 1701 | |||
AK316209.1 | - | 13 | 2048 | |||
BC130575.1 | - | 12 | 1662 | 40146606 | ||
CD364465.1 | - | 1 | 346 | |||
M16973.1 | - | 12 | 1664 | |||
X04393.1 | - | 11 | 1499 |