Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308045.1 | - | 1 | 114 | ||
BC020721.1 | - | 1 | 114 | 4734153 | ||
DB184596.1 | - | 1 | 121 | |||
X02176.1 | - | 1 | 29 |