Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127392.2 | NP_001120864.1 | 25 | 3346 | ||
NM_013279.3 | NP_037411.1 | 24 | 3153 | |||
XM_005274222.1 | XP_005274279.1 | 25 | 3337 | |||
XM_005274223.1 | XP_005274280.1 | 25 | 3322 | |||
XM_005274224.1 | XP_005274281.1 | 25 | 3319 | |||
XM_005274225.1 | XP_005274282.1 | 24 | 3256 | |||
XM_005274226.1 | XP_005274283.1 | 24 | 3253 | |||
XM_005274227.1 | XP_005274284.1 | 24 | 3241 | |||
XM_005274228.1 | XP_005274285.1 | 25 | 3184 | |||
XM_011545234.2 | XP_011543536.1 | 25 | 3223 | |||
GenBank mRNA | AB023171.2 | - | 25 | 3334 | ||
AF086762.1 | - | 24 | 3153 | |||
AK295013.1 | - | 13 | 1560 | |||
AK311259.1 | - | 2 | 929 |