Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005892.3 | NP_005883.2 | 25 | 3356 | ||
XM_006722062.3 | XP_006722125.1 | 26 | 3571 | |||
XM_006722063.3 | XP_006722126.1 | 25 | 3551 | |||
XM_006722064.3 | XP_006722127.1 | 26 | 3541 | |||
XM_006722065.3 | XP_006722128.1 | 25 | 3521 | |||
XM_006722066.3 | XP_006722129.1 | 26 | 3567 | |||
XM_006722069.2 | XP_006722132.1 | 25 | 3468 | |||
XM_006722070.3 | XP_006722133.1 | 25 | 3547 | |||
XM_011525179.2 | XP_011523481.1 | 26 | 3571 | |||
XM_011525180.2 | XP_011523482.1 | 26 | 3263 | |||
XM_011525182.2 | XP_011523484.1 | 26 | 3568 | |||
GenBank mRNA | AF432213.1 | - | 22 | 2830 | ||
AJ008112.1 | - | 10 | 1272 | |||
AK131091.1 | - | 16 | 4043 | |||
AY278319.1 | - | 25 | 3356 | |||
BC001710.1 | - | 10 | 1260 | |||
BC009000.2 | - | 5 | 537 | |||
BC021906.1 | - | 13 | 1864 | |||
BC073988.1 | - | 13 | 1904 | |||
BC111564.1 | - | 18 | 2182 | 40008718 | ||
CR456759.1 | - | 10 | 1233 | |||
FJ534522.1 | - | 9 | 1155 |