Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006134.6 | NP_006125.2 | 2 | 247 | ||
NR_040016.1 | 2 | 247 | ||||
XM_011529746.1 | XP_011528048.1 | 2 | 245 | |||
GenBank mRNA | AF045606.2 | - | 2 | 192 | ||
AF086280.1 | - | 1 | 81 | |||
AK309977.1 | - | 2 | 174 | |||
AK310701.1 | - | 2 | 103 | |||
AK311870.1 | - | 1 | 67 | |||
AY358634.1 | - | 2 | 171 | |||
AY820138.1 | - | 1 | 34 | |||
BC000569.2 | - | 2 | 163 | 3342648 | ||
BM152475.1 | - | 2 | 213 | |||
DA286739.1 | - | 2 | 248 | |||
DQ286229.1 | - | 1 | 34 | |||
DQ656043.2 | - | 2 | 249 |