Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005181.3 | NP_005172.1 | 2 | 253 | ||
GenBank mRNA | AK096880.1 | - | 2 | 253 | ||
AK300677.1 | - | 174 | ||||
AK313254.1 | - | 2 | 222 | |||
BC004897.2 | - | 2 | 208 | 3532715 | ||
DA421884.1 | - | 2 | 253 | |||
EU794596.1 | - | 2 | 853 |