Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005257639.2 | XP_005257696.1 | 8 | 963 | ||
XM_011525183.2 | XP_011523485.1 | 8 | 952 | |||
XM_017025012.1 | XP_016880501.1 | 8 | 880 | |||
GenBank mRNA | AI990988.1 | - | 1 | 271 | ||
AK289715.1 | - | 8 | 890 | |||
BC057792.1 | - | 8 | 896 | 30331755 | ||
BC069649.1 | - | 8 | 843 | 7262441 | ||
BC074768.2 | - | 8 | 820 | 30915189 | ||
CR541766.1 | - | 8 | 797 | |||
M83670.1 | - | 8 | 844 |