Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001218.4 | NP_001209.1 | 1, 2 | 476 | ||
NM_001293642.1 | NP_001280571.1 | 1, 2 | 476 | |||
NM_206925.2 | NP_996808.1 | 1, 2 | 476 | |||
NR_135511.1 | 1, 2 | 532 | ||||
GenBank mRNA | AF037335.1 | - | 1, 2 | 201 | ||
AF051882.1 | - | 1, 2 | 93 | |||
AK096845.1 | - | 1, 2 | 212 | |||
AK315769.1 | - | 1, 2 | 242 | |||
BC000278.1 | - | 1, 2 | 198 | 3357439 | ||
BC011691.2 | - | 1, 2 | 201 | 3344734 | ||
BC023981.1 | - | 1, 2 | 216 | 4042589 | ||
BQ440047.1 | - | 1, 2 | 460 | |||
BT006656.1 | - | 1, 2 | 86 | |||
DA509400.1 | - | 1, 2 | 476 | |||
DA546357.1 | - | 1, 2 | 411 |