Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000068.3 | NP_000059.3 | 36, 37 | 5648 | ||
NM_001127221.1 | NP_001120693.1 | 35, 36 | 5633 | |||
NM_001127222.1 | NP_001120694.1 | 35, 36 | 5630 | |||
NM_001174080.1 | NP_001167551.1 | 36, 37 | 5639 | |||
NM_023035.2 | NP_075461.2 | 36, 37 | 5648 | |||
GenBank mRNA | AB035726.1 | - | 36, 37 | 5432 | ||
AB035727.2 | - | 35, 36 | 5394 | |||
AF004883.1 | - | 36, 37 | 5648 | |||
AF004884.1 | - | 36, 37 | 5648 | |||
AF100774.1 | - | 8, 9 | 855 | |||
FJ040507.1 | - | 36, 37 | 5403 | |||
S76537.1 | - | 3, 4 | 321 | |||
U79663.1 | - | 12, 13 | 1463 | |||
U79664.1 | - | 12, 13 | 1463 | |||
U79665.1 | - | 12, 13 | 1463 | |||
U79666.1 | - | 35, 36 | 5614 | |||
U79667.1 | - | 12, 13 | 1463 | |||
U79668.1 | - | 12, 13 | 1463 | |||
X99897.1 | - | 35, 36 | 5675 |