Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000720.3 | NP_000711.1 | 35 | 4314 | ||
NM_001128839.2 | NP_001122311.1 | 33 | 4209 | |||
NM_001128840.2 | NP_001122312.1 | 34 | 4254 | |||
XM_005265448.3 | XP_005265505.1 | 33 | 4091 | |||
XM_011534094.2 | XP_011532396.1 | 36 | 4406 | |||
XM_011534096.2 | XP_011532398.1 | 33 | 4217 | |||
XM_011534097.2 | XP_011532399.1 | 34 | 4873 | |||
XM_011534099.2 | XP_011532401.1 | 32 | 3856 | |||
XM_011534100.2 | XP_011532402.1 | 34 | 4301 | |||
XM_017007137.1 | XP_016862626.1 | 36 | 4406 | |||
XM_017007138.1 | XP_016862627.1 | 36 | 4403 | |||
XM_017007139.1 | XP_016862628.1 | 36 | 4406 | |||
XM_017007140.1 | XP_016862629.1 | 35 | 4346 | |||
XM_017007141.1 | XP_016862630.1 | 35 | 4346 | |||
XM_017007142.1 | XP_016862631.1 | 35 | 4322 | |||
XM_017007143.1 | XP_016862632.1 | 35 | 4322 | |||
XM_017007144.1 | XP_016862633.1 | 35 | 4322 | |||
XM_017007145.1 | XP_016862634.1 | 34 | 4277 | |||
GenBank mRNA | AB209171.1 | - | 29 | 3215 | ||
AK294585.1 | - | 8 | 844 | |||
EU363339.1 | - | 33 | 4091 | |||
M76558.1 | - | 34 | 4646 | |||
M83566.1 | - | 35 | 4314 |