Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000726.3 | NP_000717.2 | 14 | 1609 | ||
NM_001005746.2 | NP_001005746.1 | 14 | 1559 | |||
NM_001005747.2 | NP_001005747.1 | 13 | 1697 | |||
NM_001145798.1 | NP_001139270.1 | 13 | 1423 | |||
NM_001320722.1 | NP_001307651.1 | 14 | 1857 | |||
XM_011511795.2 | XP_011510097.1 | 14 | 3326 | |||
XM_011511796.2 | XP_011510098.1 | 13 | 2280 | |||
XM_011511797.2 | XP_011510099.1 | 11 | 2329 | |||
XM_017004880.1 | XP_016860369.1 | 11 | 1421 | |||
XM_017004881.1 | XP_016860370.1 | 10 | 1489 | |||
XM_017004882.1 | XP_016860371.1 | 11 | 2814 | |||
XM_017004883.1 | XP_016860372.1 | 15 | 3148 | |||
XM_017004884.1 | XP_016860373.1 | 13 | 2982 | |||
XM_017004885.1 | XP_016860374.1 | 12 | 2822 | |||
XM_017004886.1 | XP_016860375.1 | 5 | 902 | |||
GenBank mRNA | AB302276.1 | - | 13 | 1367 | ||
AF038852.1 | - | 14 | 1519 | |||
AK290049.1 | - | 13 | 1678 | |||
AK291378.1 | - | 15 | 1692 | |||
AK294398.1 | - | 14 | 1533 | |||
AK316045.1 | - | 14 | 1550 | |||
AY054985.1 | - | 13 | 1668 | |||
BC075049.2 | - | 14 | 1560 | 30915578 | ||
U95020.1 | - | 14 | 1609 |