Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004929.3 | NP_004920.1 | 11 | 899 | ||
GenBank mRNA | AK054881.1 | - | 11 | 897 | ||
AK295585.1 | - | 10 | 618 | |||
AK312491.1 | - | 11 | 897 | |||
AK315959.1 | - | 10 | 605 | |||
BC006478.1 | - | 11 | 887 | 2822391 | ||
BC020864.1 | - | 11 | 897 | 4592497 | ||
BX537530.1 | - | 3 | 3756 | |||
X06661.1 | - | 11 | 738 |