Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004342.6 | NP_004333.1 | 11 | 1824 | ||
NM_033138.3 | NP_149129.2 | 12 | 2589 | |||
NM_033139.3 | NP_149130.1 | 10 | 1639 | |||
NM_033140.3 | NP_149131.1 | 9 | 1561 | |||
NM_033157.3 | NP_149347.2 | 12 | 1899 | |||
XM_017012650.1 | XP_016868139.1 | 10 | 2121 | |||
XM_017012651.1 | XP_016868140.1 | 10 | 1437 | |||
XM_017012652.1 | XP_016868141.1 | 10 | 1598 | |||
XM_017012653.1 | XP_016868142.1 | 9 | 1356 | |||
XM_017012654.1 | XP_016868143.1 | 9 | 1558 | |||
GenBank mRNA | AF247820.3 | - | 11 | 1587 | ||
AF315788.1 | - | 1 | 135 | |||
AK289686.1 | - | 12 | 1892 | |||
AK298524.1 | - | 11 | 1450 | |||
AK300282.1 | - | 9 | 1530 | |||
AK304729.1 | - | 11 | 1371 | |||
BC040354.1 | - | 11 | 1559 | 4753285 | ||
BX538339.1 | - | 1 | 63 | |||
BX648808.1 | - | 9 | 1519 | |||
D90452.1 | - | 10 | 1429 | |||
D90453.1 | - | 9 | 1351 | |||
M64110.1 | - | 10 | 1469 | |||
M83216.1 | - | 12 | 2352 |