Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004342.6 | NP_004333.1 | 11, 12 | 1901 | ||
NM_033138.3 | NP_149129.2 | 12, 13 | 2666 | |||
NM_033139.3 | NP_149130.1 | 10, 11 | 1716 | |||
NM_033140.3 | NP_149131.1 | 9, 10 | 1638 | |||
NM_033157.3 | NP_149347.2 | 12, 13 | 1976 | |||
XM_017012650.1 | XP_016868139.1 | 10, 11 | 2198 | |||
XM_017012651.1 | XP_016868140.1 | 10, 11 | 1514 | |||
XM_017012652.1 | XP_016868141.1 | 10, 11 | 1675 | |||
XM_017012653.1 | XP_016868142.1 | 9, 10 | 1433 | |||
XM_017012654.1 | XP_016868143.1 | 9, 10 | 1635 | |||
GenBank mRNA | AK289686.1 | - | 12, 13 | 1969 | ||
AK298524.1 | - | 11, 12 | 1527 | |||
AK300282.1 | - | 9, 10 | 1607 | |||
AK304729.1 | - | 11, 12 | 1448 | |||
BC040354.1 | - | 11, 12 | 1636 | 4753285 | ||
BQ010124.1 | - | 3, 4 | 710 | |||
BX538339.1 | - | 1, 2 | 140 | |||
BX648808.1 | - | 9, 10 | 1596 | |||
BX649088.1 | - | 1, 2 | 79 | |||
D90452.1 | - | 10, 11 | 1506 | |||
D90453.1 | - | 9, 10 | 1428 | |||
M64110.1 | - | 10, 11 | 1546 | |||
M83216.1 | - | 12, 13 | 2429 |