Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323374.1 | NP_001310303.1 | 6 | 523 | ||
NM_001323375.1 | NP_001310304.1 | 6 | 555 | |||
NM_001323376.1 | NP_001310305.1 | 3 | 525 | |||
NM_001323377.1 | NP_001310306.1 | 4 | 283 | |||
NM_001744.5 | NP_001735.1 | 5 | 671 | |||
GenBank mRNA | AK311662.1 | - | 3 | 380 | ||
AK312840.1 | - | 5 | 526 | |||
BC016695.1 | - | 5 | 515 | |||
BC025687.1 | - | 5 | 513 | 5205478 | ||
BG721313.1 | - | 5 | 530 | |||
D30742.1 | - | 5 | 501 | |||
L17000.1 | - | 5 | 465 | |||
L24959.1 | - | 6 | 523 |