Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CANX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024649.1 | NP_001019820.1 | 3, 4 | 362 | ||
NM_001746.3 | NP_001737.1 | 3, 4 | 415 | |||
XM_011534664.1 | XP_011532966.1 | 3, 4 | 780 | |||
XM_011534665.2 | XP_011532967.1 | 3, 4 | 388 | |||
GenBank mRNA | AJ271880.1 | - | 2, 3 | 254 | ||
AK294702.1 | - | 3, 4 | 418 | |||
AK312334.1 | - | 3, 4 | 322 | |||
BC003552.1 | - | 3, 4 | 349 | 3546389 | ||
BC042843.1 | - | 3, 4 | 296 | 4342723 | ||
L10284.1 | - | 3, 4 | 327 | |||
L18887.1 | - | 3, 4 | 333 | |||
M94859.1 | - | 3, 4 | 333 | |||
S67659.1 | - | 2, 3 | 193 |