Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256139.1 | NP_001243068.1 | 5 | 574 | ||
NM_001256140.1 | NP_001243069.1 | 5 | 864 | |||
NM_001320732.1 | NP_001307661.1 | 5 | 550 | |||
NM_001320733.1 | NP_001307662.1 | 5 | 511 | |||
NM_001320734.1 | NP_001307663.1 | 5 | 864 | |||
NM_001747.3 | NP_001738.2 | 5 | 864 | |||
XM_011533122.1 | XP_011531424.1 | 5 | 684 | |||
XM_011533123.1 | XP_011531425.1 | 5 | 580 | |||
GenBank mRNA | AK130130.1 | - | 5 | 864 | ||
AK225374.1 | - | 5 | 537 | |||
AK225378.1 | - | 5 | 537 | |||
AK300507.1 | - | 5 | 456 | |||
AK309290.1 | - | 4 | 233 | |||
AK313898.1 | - | 5 | 518 | |||
BC000728.2 | - | 5 | 518 | 3505088 | ||
BC014549.1 | - | 5 | 520 | 4108934 | ||
BG762114.1 | - | 5 | 512 | |||
BG762391.1 | - | 5 | 550 | |||
BU172582.1 | - | 5 | 516 | |||
FJ224312.1 | - | 5 | 720 | |||
M94345.1 | - | 5 | 519 |