Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014289.3 | NP_055104.2 | 8 | 1315 | ||
GenBank mRNA | AF029232.1 | - | 8 | 1215 | ||
AJ000388.1 | - | 8 | 1306 | |||
AK300340.1 | - | 6 | 871 | |||
AK312701.1 | - | 8 | 1279 | |||
BC000730.2 | - | 7 | 1144 | 3503916 |