Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPZA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006135.2 | NP_006126.1 | 7, 8 | 971 | ||
XM_017002424.1 | XP_016857913.1 | 7, 8 | 715 | |||
GenBank mRNA | AK289654.1 | - | 7, 8 | 614 | ||
BC000144.1 | - | 7, 8 | 596 | 3047937 | ||
BT019364.1 | - | 7, 8 | 579 | |||
BX648738.1 | - | 7, 8 | 1262 | |||
CR407657.1 | - | 7, 8 | 579 | |||
CR541819.1 | - | 7, 8 | 579 | |||
U56637.1 | - | 7, 8 | 579 |