Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAST SIRNA Assays ›
Gene Symbol:
CAST
Gene Name:
calpastatin
Gene Aliases:
BS-17, PLACK
Chromosome Location:
Chr.5: 96662037 - 96774683 on Build GRCh38
UniGene ID:
Hs.436186
Species:
Human
siRNA ID
HSS101363
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAST
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042440.3
NP_001035905.1
20
1782
NM_001190442.1
NP_001177371.1
21
1654
NM_001284212.2
NP_001271141.1
18
1433
NM_001284213.2
NP_001271142.1
17
1340
NM_173060.3
NP_775083.1
19
1490
NR_104285.1
7
646
XM_006714696.3
XP_006714759.1
23
1796
XM_006714697.3
XP_006714760.1
22
1813
XM_006714698.3
XP_006714761.1
21
1774
XM_006714699.3
XP_006714762.1
22
1737
XM_006714700.3
XP_006714763.1
21
1756
XM_006714701.3
XP_006714764.1
21
1772
XM_006714702.3
XP_006714765.1
21
1648
XM_006714703.3
XP_006714766.1
20
1733
XM_006714704.3
XP_006714767.1
21
1681
XM_006714705.3
XP_006714768.1
20
1690
XM_006714706.3
XP_006714769.1
20
1608
XM_006714707.3
XP_006714770.1
20
1559
XM_006714708.3
XP_006714771.1
19
1519
XM_006714709.3
XP_006714772.1
19
1482
XM_006714710.3
XP_006714773.1
18
1437
XM_006714711.3
XP_006714774.1
18
1482
XM_006714712.3
XP_006714775.1
17
1429
XM_006714713.3
XP_006714776.1
7
646
XM_006714714.3
XP_006714777.1
7
646
XM_006714715.3
XP_006714778.1
8
1372
XM_011543654.2
XP_011541956.1
21
1732
XM_011543655.2
XP_011541957.1
21
1729
XM_011543656.2
XP_011541958.1
20
1676
XM_011543657.2
XP_011541959.1
20
1558
XM_011543658.2
XP_011541960.1
18
1411
XM_017009911.1
XP_016865400.1
21
1774
XM_017009912.1
XP_016865401.1
22
1785
XM_017009913.1
XP_016865402.1
22
1754
XM_017009914.1
XP_016865403.1
21
1756
XM_017009915.1
XP_016865404.1
22
1758
XM_017009916.1
XP_016865405.1
21
1772
XM_017009917.1
XP_016865406.1
21
1690
XM_017009918.1
XP_016865407.1
20
1717
XM_017009919.1
XP_016865408.1
20
1690
XM_017009920.1
XP_016865409.1
20
1600
XM_017009921.1
XP_016865410.1
20
1617
XM_017009922.1
XP_016865411.1
19
1651
XM_017009923.1
XP_016865412.1
19
1651
XM_017009924.1
XP_016865413.1
19
1633
XM_017009925.1
XP_016865414.1
19
1569
XM_017009926.1
XP_016865415.1
19
1569
XM_017009927.1
XP_016865416.1
20
1558
XM_017009928.1
XP_016865417.1
19
1557
XM_017009929.1
XP_016865418.1
19
1476
XM_017009930.1
XP_016865419.1
18
1443
XM_017009931.1
XP_016865420.1
18
1437
XM_017009932.1
XP_016865421.1
17
1500
GenBank mRNA
AB208816.1
-
21
1618
AF327443.1
-
16
1332
AK292289.1
-
7
646
AK292356.1
-
7
619
AK296919.1
-
21
1612
AK297879.1
-
19
1432
AK298541.1
-
20
1559
AK299402.1
-
20
1539
AK300624.1
-
17
1323
AK302206.1
-
8
646
AK303836.1
-
13
1013
AK311585.1
-
8
824
AK311598.1
-
8
700
AK314290.1
-
18
1435
AK315146.1
-
18
1434
AK315673.1
-
7
594
AL832174.1
-
19
1475
AL832349.1
-
19
1478
AY189741.1
-
9
645
BC013579.1
-
20
1485
3878564
BC078149.1
-
15
1068
BT009783.1
-
17
1267
BX648397.1
-
18
1384
D16217.1
-
20
1552
U31345.1
-
9
662
U38525.1
-
2
118
U58996.2
-
7
625
Back To Top
More Information
Panther Classification:
Molecular Function -
protein-binding activity modulator
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
liver development
egg activation
brain development
myoblast fusion
aging
negative regulation of endopeptidase activity
organ regeneration
myoblast differentiation
negative regulation of cell cycle arrest
cell-cell adhesion
negative regulation of type B pancreatic cell apoptotic process
protease binding
endopeptidase inhibitor activity
protein binding
calcium-dependent cysteine-type endopeptidase inhibitor activity
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-sq56x:80/10.239.161.141:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline