Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001756.3 | NP_001747.2 | 3 | 927 | ||
GenBank mRNA | AK225591.1 | - | 4 | 837 | ||
AK290821.1 | - | 3 | 912 | |||
AK313760.1 | - | 3 | 912 | |||
BC017462.1 | - | 3 | 881 | |||
BC056259.1 | - | 3 | 902 | 5810260 | ||
BC058021.1 | - | 3 | 911 | 6714173 | ||
J02943.1 | - | 2 | 858 |