Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031966.3 | NP_114172.1 | 9 | 1508 | ||
GenBank mRNA | AK289431.1 | - | 9 | 1397 | ||
AM851053.1 | - | 8 | 1429 | |||
BC006510.2 | - | 9 | 1350 | 2963100 | ||
BT020128.1 | - | 8 | 1144 | |||
M25753.1 | - | 9 | 1344 |