Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013399.1 | NP_001013417.1 | 12 | 898 | ||
NM_005190.3 | NP_005181.2 | 12 | 1120 | |||
XM_006715594.1 | XP_006715657.1 | 13 | 1374 | |||
GenBank mRNA | AB125137.1 | - | 12 | 907 | ||
AK222768.1 | - | 12 | 931 | |||
AK223247.1 | - | 12 | 921 | |||
AK291914.1 | - | 12 | 929 | |||
AK298558.1 | - | 11 | 859 | |||
BC010135.2 | - | 12 | 953 | 4106819 | ||
BC041123.1 | - | 12 | 1103 | 5759974 | ||
BC056153.1 | - | 12 | 942 | 6157106 | ||
M74091.1 | - | 12 | 921 |