Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004060.3 | NP_004051.1 | 6 | 1106 | ||
NM_199246.1 | NP_954854.1 | 7 | 1006 | |||
GenBank mRNA | AK297190.1 | - | 6 | 719 | ||
AK312913.1 | - | 6 | 1082 | |||
AK316234.1 | - | 5 | 844 | |||
BC000196.1 | - | 7 | 956 | 3351902 | ||
BC007093.2 | - | 7 | 981 | 4251051 | ||
BT007134.1 | - | 5 | 872 | |||
BX538035.1 | - | 8 | 864 | |||
D78341.1 | - | 6 | 1062 | |||
L49504.1 | - | 6 | 1083 | |||
U47413.1 | - | 6 | 1085 | |||
X77794.1 | - | 5 | 854 |