Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004060.3 | NP_004051.1 | 2 | 389 | ||
NM_199246.1 | NP_954854.1 | 3 | 289 | |||
GenBank mRNA | AK312913.1 | - | 2 | 365 | ||
BC000196.1 | - | 3 | 239 | 3351902 | ||
BC007093.2 | - | 3 | 264 | 4251051 | ||
BM479568.1 | - | 3 | 302 | |||
BT007134.1 | - | 1 | 155 | |||
D78341.1 | - | 2 | 345 | |||
L49504.1 | - | 2 | 366 | |||
U47413.1 | - | 2 | 368 | |||
U53328.1 | - | 1 | 168 | |||
X77794.1 | - | 1 | 137 |