Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006137.6 | NP_006128.1 | 4 | 736 | ||
XM_011523608.1 | XP_011521910.1 | 4 | 703 | |||
XM_017025316.1 | XP_016880805.1 | 4 | 769 | |||
GenBank mRNA | AK223465.1 | - | 4 | 708 | ||
AK310558.1 | - | 3 | 1480 | |||
AY935534.1 | - | 4 | 708 | |||
AY935535.1 | - | 4 | 708 | |||
AY935536.1 | - | 4 | 708 | |||
AY935537.1 | - | 4 | 696 | |||
BC009293.2 | - | 4 | 680 | 4120480 | ||
BC013297.2 | - | 4 | 680 | 3529600 | ||
BI838777.1 | - | 4 | 738 | |||
BT006696.1 | - | 4 | 627 | |||
X06180.1 | - | 3 | 1114 |