Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185099.1 | NP_001172028.1 | 4 | 591 | ||
NM_001185100.1 | NP_001172029.1 | 4 | 591 | |||
NM_001185101.1 | NP_001172030.1 | 4 | 591 | |||
NM_001771.3 | NP_001762.2 | 4 | 591 | |||
GenBank mRNA | AK026467.1 | - | 2 | 1416 | ||
AK225625.1 | - | 4 | 573 | |||
AK225694.1 | - | 4 | 573 | |||
X52785.1 | - | 3 | 541 | |||
X59350.1 | - | 4 | 563 |