Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185099.1 | NP_001172028.1 | 5 | 945 | ||
NM_001185100.1 | NP_001172029.1 | 5 | 945 | |||
NM_001278417.1 | NP_001265346.1 | 4 | 773 | |||
NM_001771.3 | NP_001762.2 | 5 | 945 | |||
GenBank mRNA | AK225625.1 | - | 5 | 927 | ||
AK225694.1 | - | 5 | 927 | |||
AK301177.1 | - | 5 | 808 | |||
BC109306.1 | - | 4 | 727 | 40035425 | ||
X59350.1 | - | 5 | 917 |