Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005191.3 | NP_005182.1 | 4 | 940 | ||
XM_011513327.2 | XP_011511629.1 | 4 | 3006 | |||
XM_017007520.1 | XP_016863009.1 | 4 | 1015 | |||
GenBank mRNA | AK292129.1 | - | 4 | 924 | ||
AY197777.1 | - | 3 | 545 | |||
BC042665.1 | - | 4 | 908 | 5181343 | ||
M27533.1 | - | 4 | 862 |