Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206924.1 | NP_001193853.1 | 4 | 528 | ||
NM_001206925.1 | NP_001193854.1 | 4 | 814 | |||
NM_006889.4 | NP_008820.3 | 5 | 919 | |||
NM_175862.4 | NP_787058.4 | 5 | 864 | |||
GenBank mRNA | AK291497.1 | - | 5 | 772 | ||
AK294663.1 | - | 4 | 524 | |||
AK301237.1 | - | 4 | 810 | |||
AK313103.1 | - | 5 | 773 | |||
AK316203.1 | - | 4 | 810 | |||
BC040261.1 | - | 5 | 852 | 5173789 | ||
CR541844.1 | - | 4 | 718 | |||
L25259.1 | - | 5 | 842 | |||
U04343.1 | - | 5 | 865 |